missing translation for 'onlineSavingsMsg'
Learn More

6-(tetrahydropyran-4-yloxy)pyridine-2-carbaldehyde, Thermo Scientific™

Product Code. 10481514
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 898289-54-8
Molecular Formula C11H13NO3
Molecular Weight (g/mol) 207.229
InChI Key SBTGZJPHUYLRRU-UHFFFAOYSA-N
Synonym 6-tetrahydropyran-4-yloxy pyridine-2-carbaldehyde, 6-oxan-4-yloxy pyridine-2-carbaldehyde, 6-oxan-4-yl oxy pyridine-2-carbaldehyde, 6-tetrahydro-2h-pyran-4-yl oxy picolinaldehyde, 6-2h-3,4,5,6-tetrahydropyran-4-yloxy pyridine-2-carbaldehyde
PubChem CID 45594294
IUPAC Name 6-(oxan-4-yloxy)pyridine-2-carbaldehyde
SMILES C1COCCC1OC2=CC=CC(=N2)C=O

Specifications

Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 207.23
Percent Purity 95%
Chemical Name or Material 6-(tetrahydropyran-4-yloxy)pyridine-2-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'