missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-thien-2-ylnicotinic acid, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
Assay Percent Range | 97% |
Quantity | 1 g |
IUPAC Name | 6-(thiophen-2-yl)pyridine-3-carboxylic acid |
Formula Weight | 205.24 |
Percent Purity | 97% |
Chemical Name or Material | 6-Thien-2-ylnicotinic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'