missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-thien-2-ylnicotinic acid, Thermo Scientific™
£59.80 - £239.00
Chemical Identifiers
CAS | 179408-54-9 |
---|---|
Molecular Formula | C10H7NO2S |
Molecular Weight (g/mol) | 205.23 |
MDL Number | MFCD06410384 |
InChI Key | QYGFXNUFKLRCRB-UHFFFAOYSA-N |
Synonym | 6-thien-2-ylnicotinic acid, 6-thiophen-2-yl nicotinic acid, 6-2-thienyl nicotinic acid, 6-thiophen-2-yl pyridine-3-carboxylic acid, 6-2-thienyl pyridine-3-carboxylic acid, acmc-209efk, 6-thien-2ylnicotinic acid, 6-thiophen-2-yl-nicotinic acid, 6-thiophen-2-ylnicotinic acid, 2-thien-2-yl-5-pyridinecarboxylic acid |
PubChem CID | 15411144 |
SMILES | OC(=O)C1=CN=C(C=C1)C1=CC=CS1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10623245
|
Thermo Scientific Maybridge
CC54801CB |
250 mg |
£59.80
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10418643
|
Thermo Scientific Maybridge
CC54801DA |
1 g |
£148.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10326384
|
Thermo Scientific Maybridge
CC54801DE |
5 g |
£239.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
179408-54-9 | |
205.23 | |
QYGFXNUFKLRCRB-UHFFFAOYSA-N | |
15411144 |
C10H7NO2S | |
MFCD06410384 | |
6-thien-2-ylnicotinic acid, 6-thiophen-2-yl nicotinic acid, 6-2-thienyl nicotinic acid, 6-thiophen-2-yl pyridine-3-carboxylic acid, 6-2-thienyl pyridine-3-carboxylic acid, acmc-209efk, 6-thien-2ylnicotinic acid, 6-thiophen-2-yl-nicotinic acid, 6-thiophen-2-ylnicotinic acid, 2-thien-2-yl-5-pyridinecarboxylic acid | |
OC(=O)C1=CN=C(C=C1)C1=CC=CS1 |
Specifications
179408-54-9 | |
C10H7NO2S | |
6-thien-2-ylnicotinic acid, 6-thiophen-2-yl nicotinic acid, 6-2-thienyl nicotinic acid, 6-thiophen-2-yl pyridine-3-carboxylic acid, 6-2-thienyl pyridine-3-carboxylic acid, acmc-209efk, 6-thien-2ylnicotinic acid, 6-thiophen-2-yl-nicotinic acid, 6-thiophen-2-ylnicotinic acid, 2-thien-2-yl-5-pyridinecarboxylic acid | |
OC(=O)C1=CN=C(C=C1)C1=CC=CS1 | |
205.23 | |
205.24 | |
6-Thien-2-ylnicotinic acid |
97% | |
MFCD06410384 | |
QYGFXNUFKLRCRB-UHFFFAOYSA-N | |
6-thiophen-2-ylpyridine-3-carboxylic acid | |
15411144 | |
97% |
Safety and Handling
GHS Signal Word: Warning