missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Thioguanine, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
Anticancer and immunosuppressive agent often used to treat immune disorders and leukemia. Displays cytotoxic and antineoplastic properties; disrupts cytosine methylation by DNA methyltransferases after incorporation into DNA. Selectively kills BRCA2-defective tumors in a xenograft model. Also facilitates proteasome-mediated degradation of DNA (cytosine-5)-methyltransferase 1 (DNMT1). Also inhibits SARS-CoV papain-like protease (PLpro; IC50 = 5 μM for PLpro deubiquitination activity).
Specifications
Specifications
CAS | 154-42-7 |
Molecular Formula | C5H5N5S |
MDL Number | MFCD00233553 |
Quantity | 50 mg |
Synonym | 6-thioguanine, thioguanine, tioguanine, 2-amino-6-mercaptopurine, 6-mercaptoguanine, tioguanin, tabloid, lanvis, 2-amino-6-purinethiol, 2-aminopurine-6-thiol |
InChI Key | WYWHKKSPHMUBEB-UHFFFAOYSA-N |
SMILES | NC1=NC(=S)C2=C(N1)N=CN2 |
IUPAC Name | 2-amino-6,7-dihydro-3H-purine-6-thione |
Molecular Weight (g/mol) | 167.19 |
PubChem CID | 2723601 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'