Learn More
Ohaus™ 6 x Centrifuge Tube Adapters, Frontier™ R-A12x5/14MS Rotors
Facilitate centrifugation of tubes smaller than the original, provided carrier space for Frontier™ R-A12x5/14MS Rotors
Brand:  Ohaus™ 30130888
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15876632
  • £133.00 / Pack of 6
In Stock
to see stock.
Description
Description
  • Allows you to centrifuge tubes that are smaller in number than the space provided in the rotor carrier
  • Suitable for use in Frontier™ R-A12x5/14MS Rotors
Specifications
Specifications
6 x 1.8 mL
R-A12x5/14MS Rotor
1
12.6 mm
Adapter, D12.6mm, 1.8ml Cryo x6
6