Learn More
YSI™ 60510 Pro Series ISE Cable
The 60510 is a 1-port cable for use with the Professional Plus instrument. The port requires a ISE probe.
Brand:  YSI™ 60510-10
 View more versions of this product
Product Code. 12963487
  • £293.00 / Each
Estimated Shipment: 26-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Conductivity Probes and Multiparameter Cables. Order other probes separately.
  • The 60510 ISE (pH, ORP, pH/ORP, ammonium, nitrate, chloride) cables ONLY work with the YSI Pro Plus handheld
  • Any of the ISE sensors can be connected to this cable at any time
    • TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cable management kit
393.7 in.
10 m