missing translation for 'onlineSavingsMsg'
Learn More

(6R)-5,6,7,8-Tetrahydrobiopterin dihydrochloride, Thermo Scientific Chemicals

£132.66 - £459.29

Chemical Identifiers

CAS 69056-38-8
Molecular Formula C9H15N5O3·2HCl
Molecular Weight (g/mol) 314.17
InChI Key RKSUYBCOVNCALL-NTVURLEBSA-N
Synonym sapropterin dihydrochloride, biopten, r-2-amino-6-1r,2s-1,2-dihydroxypropyl-5,6,7,8-tetrahydropteridin-4 3h-one dihydrochloride, kuvan, sapropterin hydrochloride, unii-rg277lf5b3, biopten tn, sapropterin hcl, 6r-tetrahydrobiopterin hydrochloride
PubChem CID 636369
ChEBI CHEBI:32120
IUPAC Name (6R)-2-amino-6-[(1R,2S)-1,2-dihydroxypropyl]-5,6,7,8-tetrahydro-1H-pteridin-4-one;dihydrochloride
SMILES CC(C(C1CNC2=C(N1)C(=O)N=C(N2)N)O)O.Cl.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15743447
View Documents
Thermo Scientific Acros
328871000
100 mg
£132.66
100mg
Please to purchase this item. Need a web account? Register with us today!
15753447
View Documents
Thermo Scientific Acros
328875000
500 mg
£459.29
500mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

69056-38-8
314.17
sapropterin dihydrochloride, biopten, r-2-amino-6-1r,2s-1,2-dihydroxypropyl-5,6,7,8-tetrahydropteridin-4 3h-one dihydrochloride, kuvan, sapropterin hydrochloride, unii-rg277lf5b3, biopten tn, sapropterin hcl, 6r-tetrahydrobiopterin hydrochloride
CHEBI:32120
CC(C(C1CNC2=C(N1)C(=O)N=C(N2)N)O)O.Cl.Cl
C9H15N5O3·2HCl
RKSUYBCOVNCALL-NTVURLEBSA-N
636369
(6R)-2-amino-6-[(1R,2S)-1,2-dihydroxypropyl]-5,6,7,8-tetrahydro-1H-pteridin-4-one;dihydrochloride

Specifications

69056-38-8
C9H15N5O3·2HCl
sapropterin dihydrochloride, biopten, r-2-amino-6-1r,2s-1,2-dihydroxypropyl-5,6,7,8-tetrahydropteridin-4 3h-one dihydrochloride, kuvan, sapropterin hydrochloride, unii-rg277lf5b3, biopten tn, sapropterin hcl, 6r-tetrahydrobiopterin hydrochloride
RKSUYBCOVNCALL-NTVURLEBSA-N
(6R)-2-amino-6-[(1R,2S)-1,2-dihydroxypropyl]-5,6,7,8-tetrahydro-1H-pteridin-4-one;dihydrochloride
636369
314.17
250.0°C to 255.0°C
15,8505
Solubility in water: soluble.
CC(C(C1CNC2=C(N1)C(=O)N=C(N2)N)O)O.Cl.Cl
314.17
CHEBI:32120
(6R)-5,6,7,8-Tetrahydrobiopterin dihydrochloride
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only