missing translation for 'onlineSavingsMsg'
Learn More

6x-His Tag Monoclonal Antibody (4E3D10H2/E3), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA1135

Code : NEW

Additional Details : Weight : 0.00001kg

Product Code. 15442890

  • £229.00 / 100µg
Estimated Shipment: 30-07-2024
to see stock.

Description

Description

By Western blot, MA1-135 detects both N-terminal and C-terminal His-tagged proteins.

6x-His Tag Monoclonal antibody specifically detects 6x-His Tag in Tag samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot.
Specifications

Specifications

6x-His Tag
Monoclonal
1 mg/mL
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide
Mouse
Protein A
RUO
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Tag
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
4E3D10H2/E3
Unconjugated
6His tag; 6X His; His Tag; Histidine Tag
Mixture of N-terminal and C-terminal 6x-His peptides, CG-HHHHHH and HHHHHH-GC
100 μg
Primary
Antibody
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers