missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydrobenzo[b]furan, 95%, Thermo Scientific™
£59.80 - £155.00
Chemical Identifiers
CAS | 934586-50-2 |
---|---|
Molecular Formula | C14H19BO3 |
Molecular Weight (g/mol) | 246.11 |
MDL Number | MFCD11109314 |
InChI Key | HCAAUTYLSSDHFV-UHFFFAOYSA-N |
Synonym | 2-2,3-dihydrobenzofuran-7-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydrobenzo b furan, 2,3-dihydrobenzofuran-7-boronic acid, pinacol ester, 2-2,3-dihydro-1-benzofuran-7-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 7-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2,3-dihydro-benzofuran, 7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1-benzofuran, 2,3-dihydrobenzo b furan-7-boronic acid pinacol ester, 2-2,3-dihydrobenzo 2,3-b furan-7-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
PubChem CID | 43811027 |
IUPAC Name | 2-(2,3-dihydro-1-benzofuran-7-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
SMILES | CC1(C)OB(OC1(C)C)C1=C2OCCC2=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10642655
|
Thermo Scientific Maybridge
CC00939CB |
250 mg | Amber Glass Bottle |
£59.80
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10652655
|
Thermo Scientific Maybridge
CC00939DA |
1 g | Amber Glass Bottle |
£155.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
934586-50-2 | |
246.11 | |
HCAAUTYLSSDHFV-UHFFFAOYSA-N | |
43811027 | |
CC1(C)OB(OC1(C)C)C1=C2OCCC2=CC=C1 |
C14H19BO3 | |
MFCD11109314 | |
2-2,3-dihydrobenzofuran-7-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydrobenzo b furan, 2,3-dihydrobenzofuran-7-boronic acid, pinacol ester, 2-2,3-dihydro-1-benzofuran-7-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 7-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2,3-dihydro-benzofuran, 7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1-benzofuran, 2,3-dihydrobenzo b furan-7-boronic acid pinacol ester, 2-2,3-dihydrobenzo 2,3-b furan-7-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
2-(2,3-dihydro-1-benzofuran-7-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Specifications
934586-50-2 | |
100.0 | |
C14H19BO3 | |
2-2,3-dihydrobenzofuran-7-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydrobenzo b furan, 2,3-dihydrobenzofuran-7-boronic acid, pinacol ester, 2-2,3-dihydro-1-benzofuran-7-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 7-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2,3-dihydro-benzofuran, 7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1-benzofuran, 2,3-dihydrobenzo b furan-7-boronic acid pinacol ester, 2-2,3-dihydrobenzo 2,3-b furan-7-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
CC1(C)OB(OC1(C)C)C1=C2OCCC2=CC=C1 | |
246.11 | |
246.11 | |
7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydrobenzo[b]furan |
94.5 | |
95% | |
MFCD11109314 | |
HCAAUTYLSSDHFV-UHFFFAOYSA-N | |
2-(2,3-dihydro-1-benzofuran-7-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
43811027 | |
95% |
Safety and Handling
Warning