missing translation for 'onlineSavingsMsg'
Learn More

7-Amino-1,3-naphthalenedisulfonic acid, Tech., Thermo Scientific Chemicals

Product Code. 10219873
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-65-7
Molecular Formula C10H9NO6S2
Molecular Weight (g/mol) 303.29
MDL Number MFCD00003991
InChI Key CMOLPZZVECHXKN-UHFFFAOYSA-N
Synonym amido-g-acid, 7-amino-1,3-naphthalenedisulfonic acid, amino-g-acid, 1,3-naphthalenedisulfonic acid, 7-amino, amino-g acid, 2-amino-6,8-disulfonaphthalene, unii-7c482fx29k, 2-aminonaphthalene-6,8-disulfonic acid, 2-naphthylamine-6,8-disulfonic acid, beta-naphthylamine-6,8-disulfonic acid
PubChem CID 6851
IUPAC Name 7-aminonaphthalene-1,3-disulfonic acid
SMILES C1=CC(=CC2=C(C=C(C=C21)S(=O)(=O)O)S(=O)(=O)O)N

Specifications

Melting Point >300°C
Color Brown to White
Infrared Spectrum Authentic
Packaging Plastic Bottle
Quantity 2.5 kg
Beilstein 14, 784
Merck Index 15, 395
Solubility Information Solubility in water: soluble
Formula Weight 303.29
Percent Purity 60% (as is basis) Typically
Grade Technical
Physical Form Needles or Powder
Chemical Name or Material 7-Amino-1, 3-naphthalenedisulfonic acid, tech.
Show More Show Less
compliance-icons
Product Identifier
  • 7-Amino-1,3-naphthalenedisulfonic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'