Learn More
Molecular Probes™ 7-Amino-4-Methylcoumarin, reference standard
7-Amino-4-methylcoumarin (AMC) can be used as a fluorescence reference standard for AMC-based enzyme substrates including AMC-based caspase substrates.
Brand: Molecular Probes™ A191
97.33 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
7-Amino-4-methylcoumarin (AMC) can be used as a fluorescence reference standard for AMC-based enzyme substrates including AMC-based caspase substrates. The fluorescence spectra of AMC is not subject to variability due to pH-dependent protonation/deprotonation when assayed near or above physiological pH.
To make a concentrated stock solution of up to 10 mg/mL, dissolve in either acetone, high quality anhydrous DMSO or DMF. Store frozen, desiccated and protected from light.
Specifications
100 mg | |
175.2 | |
Room Temperature | |
Coumarin Dyes | |
351 nm |
7-Amino-4-Methylcoumarin | |
Store at room temperature and protect from light. | |
Methylcoumarin | |
430 nm |
For Research Use Only. Not for use in diagnostic procedures.