missing translation for 'onlineSavingsMsg'
Learn More
(7-Aza-1H-benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate, 99+%, Thermo Scientific Chemicals Product Code.: 15485265

(7-Aza-1H-benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate, 99+%, Thermo Scientific Chemicals

Product Code. 15485265
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15485265

Brand: Thermo Scientific Alfa Aesar H54378.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

(7-Aza-1H-benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate is used as reagent for synthesis of cyclic RGC pentapeptides for functionalization through click chemistry, fluorescent glucose bioprobes and ReactIR™ flow cell. It is also used as reagent for disulfide bond engineering, heteroaryl ether synthesis and optimization of conformational constraint on non-phosphorylated cyclic peptide antagonists of the Grb2-SH2 domain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(7-Aza-1H-benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate is used as reagent for synthesis of cyclic RGC pentapeptides for functionalization through click chemistry, fluorescent glucose bioprobes and ReactIR™ flow cell. It is also used as reagent for disulfide bond engineering, heteroaryl ether synthesis and optimization of conformational constraint on non-phosphorylated cyclic peptide antagonists of the Grb2-SH2 domain.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156311-83-0
Molecular Formula C17H27F6N7OP2
Molecular Weight (g/mol) 521.39
MDL Number MFCD03703417
InChI Key CBZAHNDHLWAZQC-UHFFFAOYSA-N
Synonym pyaop, 3h-1,2,3 triazolo 4,5-b pyridin-3-yl oxy tri pyrrolidin-1-yl phosphonium hexafluorophosphate v, 7-azabenzotriazol-1-yloxy tripyrrolidinophosphonium hexafluorophosphate, 3-hydroxy-3h-1,2,3-triazolo 4,5-b pyridinato-o tri-1-pyrrolidinylphosphonium hexafluorophosphate, pyaop reagent, 7-azabenzotriazol-1-yloxy tripyrrolidino-phosphonium hexafluorophosphate, 7-azabenzotriazol-1-yloxy-tris-pyrrolidino phosphonium hexafluorophosphate, pubchem12746, ksc491e1h
PubChem CID 11038641
IUPAC Name tripyrrolidin-1-yl(triazolo[4,5-b]pyridin-3-yloxy)phosphanium;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.C1CCN(C1)[P+](ON1N=NC2=CC=CN=C12)(N1CCCC1)N1CCCC1

Specifications

Melting Point 163°C to 168°C
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 521.38
Percent Purity ≥99%
Chemical Name or Material (7-Aza-1H-benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX14K002 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog NumberH54378.03,H54378.06
Lot NumberW01G004 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH54378.03,H54378.06
Lot NumberBC150508 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH54378.03,H54378.06
Lot Number81401430 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH54378.03,H54378.06
Lot NumberW20H022 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH54378.03,H54378.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(7-Aza-1H-benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.