missing translation for 'onlineSavingsMsg'
Learn More

7-Aza-1H-benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate, 98+%, Thermo Scientific™

Product Code. 11352469
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11352469

Brand: Thermo Scientific Alfa Aesar H26609.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156311-85-2
Molecular Formula C11H21F6N7OP2
Molecular Weight (g/mol) 443.275
MDL Number MFCD09263289
InChI Key RQBNNDQCKMIUQJ-UHFFFAOYSA-N
Synonym 3h-1,2,3 triazolo 4,5-b pyridin-3-yl oxy tris dimethylamino phosphonium hexafluorophosphate v, 7-azabenzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, 7-azabenzotriazol-1-yloxytris dimethylamino phosph, 7-aza-1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, acmc-20alxg, tris dimethylamino 3h-1,2,3-triazolo 4,5-b pyridin-3-yloxy phosphonium hexafluorophosphate, ksc173q2f, 7-azabenzotriazol-1-yloxytris dimethyamino phosphonium hexafluorosphate, 7-azabenzotriazol-1-yloxy tris dimethylamino phosphonium hexafluorophosphate, 7-aza-1h-benzotriazol-1-yloxytris dimethylamino phosphonium hexafluorophosphate, 98+%
PubChem CID 21894514
IUPAC Name tris(dimethylamino)-(triazolo[4,5-b]pyridin-3-yloxy)phosphanium;hexafluorophosphate
SMILES CN(C)[P+](N(C)C)(N(C)C)ON1C2=C(C=CC=N2)N=N1.F[P-](F)(F)(F)(F)F

Specifications

Melting Point ∽180°C (decomposition)
Quantity 1 g
Formula Weight 443.27
Percent Purity ≥98%
Chemical Name or Material 7-Aza-1H-benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.