missing translation for 'onlineSavingsMsg'
Learn More
(7-Azabenzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate, 98%, Thermo Scientific Chemicals Product Code.: 11966701

(7-Azabenzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate, 98%, Thermo Scientific Chemicals

Product Code. 11966701
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 11966701

Brand: Thermo Scientific Acros 440610050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156311-83-0
Molecular Formula C17H27F6N7OP2
Molecular Weight (g/mol) 521.39
MDL Number MFCD03703417
InChI Key CBZAHNDHLWAZQC-UHFFFAOYSA-N
Synonym pyaop, 3h-1,2,3 triazolo 4,5-b pyridin-3-yl oxy tri pyrrolidin-1-yl phosphonium hexafluorophosphate v, 7-azabenzotriazol-1-yloxy tripyrrolidinophosphonium hexafluorophosphate, 3-hydroxy-3h-1,2,3-triazolo 4,5-b pyridinato-o tri-1-pyrrolidinylphosphonium hexafluorophosphate, pyaop reagent, 7-azabenzotriazol-1-yloxy tripyrrolidino-phosphonium hexafluorophosphate, 7-azabenzotriazol-1-yloxy-tris-pyrrolidino phosphonium hexafluorophosphate, pubchem12746, ksc491e1hShow MoreShow Less
PubChem CID 11038641
IUPAC Name tripyrrolidin-1-yl(triazolo[4,5-b]pyridin-3-yloxy)phosphanium;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.C1CCN(C1)[P+](ON1N=NC2=CC=CN=C12)(N1CCCC1)N1CCCC1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 153°C to 155°C
Color White
Elemental Analysis 38.18 to 40.14% C, 18.33 to 19.28% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Formula Weight 521.39
Percent Purity 98%
Physical Form Crystalline Powder or Powder
Chemical Name or Material (7-Azabenzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number344689 Certificate TypeCertificate of Analysis Date30/09/2024 Catalog Number440610010,440610050
Lot Number343470 Certificate TypeCertificate of Analysis Date17/01/2023 Catalog Number440610010,440610050
Lot Number344467 Certificate TypeCertificate of Analysis Date11/01/2023 Catalog Number440610010,440610050
Lot Number344281 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number440610010,440610050
Lot Number344689 Certificate TypeCertificate of Analysis Date22/09/2021 Catalog Number
compliance-icons
Product Identifier
  • PyAOP
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(7-Azabenzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.