missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-1-chloroisoquinoline, 97%, Thermo Scientific Chemicals

Product Code. 11349955
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

7-Bromo-1-chloroisoquinoline as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Bromo-1-chloroisoquinoline as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 215453-51-3
Molecular Formula C9H5BrClN
Molecular Weight (g/mol) 242.5
MDL Number MFCD04039320
InChI Key UMSWWSIVPWVJOX-UHFFFAOYSA-N
Synonym 7-bromo-1-chloro-isoquinoline, 1-chloro-7-bromoisoquinoline, isoquinoline, 7-bromo-1-chloro, acmc-209y3w
PubChem CID 6502024
IUPAC Name 7-bromo-1-chloroisoquinoline
SMILES C1=CC(=CC2=C1C=CN=C2Cl)Br

Specifications

Melting Point 122°C to 126°C
Quantity 1 g
Beilstein 8404252
Solubility Information Insoluble in water.
Formula Weight 242.5
Percent Purity 97%
Chemical Name or Material 7-Bromo-1-chloroisoquinoline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.