missing translation for 'onlineSavingsMsg'
Learn More
7-Bromo-3,4-dihydro-2H-1,4-benzothiazine, 97%, Thermo Scientific Chemicals Product Code.: 15487508

7-Bromo-3,4-dihydro-2H-1,4-benzothiazine, 97%, Thermo Scientific Chemicals

Product Code. 15487508
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15487508

Brand: Thermo Scientific Alfa Aesar H61021.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

7-Bromo-3,4-dihydro-2H-1,4-benzothiazine used in many areas of medicine. They are used in industries as dyes and as antioxidants of rubber and natural elastomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Bromo-3,4-dihydro-2H-1,4-benzothiazine used in many areas of medicine. They are used in industries as dyes and as antioxidants of rubber and natural elastomers.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 193414-60-7
Molecular Formula C8H8BrNS
Molecular Weight (g/mol) 230.12
MDL Number MFCD11847754
InChI Key QWWYGAWGUCQKDL-UHFFFAOYSA-N
Synonym 7-bromo-3,4-dihydro-2h-benzo b 1,4 thiazine
PubChem CID 83823811
IUPAC Name 7-bromo-3,4-dihydro-2H-1,4-benzothiazine
SMILES BrC1=CC2=C(NCCS2)C=C1

Specifications

Quantity 1 g
Formula Weight 230.12
Percent Purity 97%
Chemical Name or Material 7-Bromo-3,4-dihydro-2H-1,4-benzothiazine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberH16Z047 Certificate TypeCertificate of Analysis Date18/01/2025 Catalog NumberH61021.03
Lot NumberM18K011 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog NumberH61021.03,H61021.MD
Lot NumberZ21J015 Certificate TypeCertificate of Analysis Date10/01/2024 Catalog NumberH61021.03,H61021.MD
Lot NumberY02J005 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog NumberH61021.03,H61021.MD
Lot NumberH16Z047 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH61021.03
compliance-icons
Product Identifier
  • 7-Bromo-3,4-dihydro-2H-1,4-benzothiazine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 7-Bromo-3,4-dihydro-2H-1,4-benzothiazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
7-Bromo-3,4-dihydro-2H-1,4-benzothiazine, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.