missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Bromo-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
IUPAC Name | 7-bromo-2H,3H,4H-pyrido[3,2-b][1,4]oxazine |
Formula Weight | 215.05 |
Percent Purity | 97% |
Chemical Name or Material | 7-Bromo-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
7-Bromo-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction