missing translation for 'onlineSavingsMsg'
Learn More

7-Bromoindole, 98%, Thermo Scientific™

Product Code. 11458620
Click to view available options
Quantity:
0.25 g
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51417-51-7
Molecular Formula C8H6BrN
Molecular Weight (g/mol) 196.047
MDL Number MFCD00799492
InChI Key RDSVSEFWZUWZHW-UHFFFAOYSA-N
Synonym 7-bromoindole, 1h-indole, 7-bromo, 7-bromo-indole, 7-bromindol, 7-bromoindol, 7-brom-1h-indole, pubchem7318, acmc-209ktm, ksc269i1f
PubChem CID 2757020
IUPAC Name 7-bromo-1H-indole
SMILES C1=CC2=C(C(=C1)Br)NC=C2

Specifications

Melting Point 42°C to 44°C
Quantity 0.25 g
Sensitivity Light sensitive
Formula Weight 196.05
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 7-Bromoindole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'