missing translation for 'onlineSavingsMsg'
Learn More

7-Deazahypoxanthine, 97%, Thermo Scientific Chemicals

Product Code. 15491799
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as pharmaceutical intermediate. Methylated 7-Deazahypoxanthines is used as regiochemical probes of xanthine oxidase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Methylated 7-Deazahypoxanthines is used as regiochemical probes of xanthine oxidase.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Moisture and light sensitive. Store at -20°C
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3680-71-5
Molecular Formula C6H5N3O
Molecular Weight (g/mol) 135.13
MDL Number MFCD08445809
InChI Key FBMZEITWVNHWJW-UHFFFAOYSA-N
Synonym 7h-pyrrolo 2,3-d pyrimidin-4-ol, pyrrolo 2,3-d pyrimidin-4-ol, 7-deazahypoxanthine, 3h-pyrrolo 2,3-d pyrimidin-4 7h-one, 4-hydroxypyrrolo 2,3-d pyrimidine, 4-hydroxypyrrolopyrimidine, 1h-pyrrolo 2,3-d pyrimidin-4 7h-one, 3,7-dihydro-4h-pyrrolo 2,3-d pyrimidin-4-one, pyrrolo 2,3-d pyrimidin-4-one
PubChem CID 96194
IUPAC Name 1,7-dihydropyrrolo[2,3-d]pyrimidin-4-one
SMILES O=C1N=CNC2=C1C=CN2

Specifications

Melting Point 345°C to 350°C
Color Brown
Quantity 1 g
Beilstein 607869
Sensitivity Light and moisture sensitive
Solubility Information Soluble in dimethyl sulfoxide.
Formula Weight 135.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 7-Deazahypoxanthine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'