missing translation for 'onlineSavingsMsg'
Learn More
7-Ethoxyresorufin, Thermo Scientific Chemicals Product Code.: 15455269

7-Ethoxyresorufin, Thermo Scientific Chemicals

Product Code. 15455269
5 mg
Click to view available options
Quantity:
5 mg
This item is not returnable. View return policy

Product Code. 15455269

Brand: Thermo Scientific Alfa Aesar J62987.#0

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

Fluorimetric substrate for detection of cytochrome P450 linked enzymes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5725-91-7
Melting Point 237°C to 240°C
Color Brown
Molecular Formula C14H11NO3
MDL Number MFCD00037661
Quantity 5 mg
Beilstein 225973
Synonym 7-ethoxyresorufin, ethoxyresorufin, resorufin ethyl ether, 7-ethoxy-3h-phenoxazin-3-one, 7-ethoxyphenoxazone, 3h-phenoxazin-3-one, 7-ethoxy, o7-ethylresorufin, 3h-phenoxazin-3-one,7-ethoxy, 7-ethoxy resorufin, 7-ethoxy-3-phenoxazinone
InChI Key CRCWUBLTFGOMDD-UHFFFAOYSA-N
SMILES CCOC1=CC2=C(C=C1)N=C3C=CC(=O)C=C3O2
IUPAC Name 7-ethoxyphenoxazin-3-one
Molecular Weight (g/mol) 241.246
PubChem CID 3294
ChEBI CHEBI:34480
Formula Weight 241.24
Physical Form Powder
Chemical Name or Material 7-Ethoxyresorufin
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX02K009 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog NumberJ62987.#0
Lot NumberQ05K001 Certificate TypeCertificate of Analysis Date17/04/2024 Catalog NumberJ62987.#0
Lot NumberT28J020 Certificate TypeCertificate of Analysis Date14/08/2023 Catalog NumberJ62987.#0
Lot NumberS20J006 Certificate TypeCertificate of Analysis Date27/06/2023 Catalog NumberJ62987,J62987.#0
Lot NumberK21Z036 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ62987.#0
compliance-icons
Product Identifier
  • 7-Ethoxyresorufin
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 7-Ethoxyresorufin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
7-Ethoxyresorufin, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.