missing translation for 'onlineSavingsMsg'
Learn More

7-Fluorobenzofurazan-4-sulfonamide, 98%, Thermo Scientific Chemicals

Product Code. 11435617
Click to view available options
Quantity:
25 mg
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91366-65-3
Molecular Formula C6H4FN3O3S
Molecular Weight (g/mol) 217.17
MDL Number MFCD00036687
InChI Key XROXHZMRDABMHS-UHFFFAOYSA-N
Synonym abd-f, 4-aminosulfonyl-7-fluoro-2,1,3-benzoxadiazole, 7-fluorobenzofurazan-4-sulfonamide, 4-fluoro-7-sulfamoylbenzofurazan, 7-fluorobenzo c 1,2,5 oxadiazole-4-sulfonamide, 4-aminosulphonyl-7-fluoro-2,1,3-benzoxadiazole, 7-fluorobenzo c 1,2,5-oxadiazole-4-sulfonamide, pubchem23230, 4-fluoro-7-sulfamoylbenzofurazane, 4-fluoro-7-sulfoamoylbenzofurazan
PubChem CID 122067
IUPAC Name 7-fluoro-2,1,3-benzoxadiazole-4-sulfonamide
SMILES NS(=O)(=O)C1=CC=C(F)C2=NON=C12

Specifications

Melting Point 143°C to 147°C
Quantity 100 mg
Formula Weight 217.17
Percent Purity 98%
Chemical Name or Material 7-Fluorobenzofurazan-4-sulfonamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.