missing translation for 'onlineSavingsMsg'
Learn More

7-Fluoroindole, 97%

Product Code. 11358566
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11358566

Brand: Thermo Scientific Alfa Aesar L17621.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 387-44-0 | C8H6FN | 135.14 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in methanol. (almost transparency)

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 387-44-0
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.14
MDL Number MFCD01074502
InChI Key XONKJZDHGCMRRF-UHFFFAOYSA-N
Synonym 7-fluoroindole, 1h-indole, 7-fluoro, pubchem7294, 7-fluoranyl-1h-indole, acmc-209j1q, ksc222e4r, 7-fluoro-1h-indole, buttpark 24\07-35
PubChem CID 2774504
IUPAC Name 7-fluoro-1H-indole
SMILES FC1=C2NC=CC2=CC=C1

Specifications

Melting Point 61°C to 63°C
Quantity 1 g
Beilstein 114740
Solubility Information Soluble in methanol. (almost transparency)
Formula Weight 135.14
Percent Purity 97%
Chemical Name or Material 7-Fluoroindole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.