missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Hydroxy-4'-nitroisoflavone, 98%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
100 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
7-Hydroxy-4′-nitroisoflavone is used in the synthesis of iso flavones, Synthesis of daidzin, soflavone amide derivatives.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
7-Hydroxy-4′-nitroisoflavone is used in the synthesis of iso flavones, Synthesis of daidzin, soflavone amide derivatives.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 290°C to 292°C |
Color | Yellow |
Quantity | 5 g |
Beilstein | 385006 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 283.24 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | 7-Hydroxy-4'-nitroisoflavone |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'