missing translation for 'onlineSavingsMsg'
Learn More

7-(Trifluoromethyl)-1,2,3,4-tetrahydroquinoline, 97%

Product Code. p-7045289
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 11328265

Brand: Thermo Scientific Alfa Aesar L10150.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 450-62-4 | C10H10F3N | 201.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 450-62-4
Molecular Formula C10H10F3N
Molecular Weight (g/mol) 201.192
MDL Number MFCD00079784
InChI Key RGZZKZNESVFQKR-UHFFFAOYSA-N
Synonym 7-trifluoromethyl-1,2,3,4-tetrahydroquinoline, 7-trifluoromethyl-1,2,3,4-tetrahydro-quinoline, quinoline, 1,2,3,4-tetrahydro-7-trifluoromethyl, pubchem7221, acmc-20aip4
PubChem CID 2781139
IUPAC Name 7-(trifluoromethyl)-1,2,3,4-tetrahydroquinoline
SMILES C1CC2=C(C=C(C=C2)C(F)(F)F)NC1

Specifications

Melting Point 30°C to 34°C
Boiling Point 135°C to 140°C (22 mmHg)
Quantity 250 mg
Beilstein 153990
Formula Weight 201.19
Percent Purity 97%
Chemical Name or Material 7-(Trifluoromethyl)-1,2,3,4-tetrahydroquinoline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.