missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ 70-215 Series Yellow Neptune™ Medium Weight Gloves

Product Code. p-8004580 Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 12670972

Brand: Ansell™ 70215080

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Constructed from high-tech para-aramid short fibers, the gloves provide improved sensitivity and fit. The seamless construction avoids irritation and the use of short fibers offer extra comfort.

  • Industrial gloves
  • 100% DuPont™ Kevlar™ Aramid fibers glove provides light-duty cut protection
  • Protection from sharp edges of metals, ceramics, glass and other materials
  • Reversible design allows glove to be worn on either hand, reducing replacement costs
  • Offers very high cut resistance to weight ratio
  • Use of Terry loop technique enhances heat insulation
  • Thicker-walled glove offers high shock absorption without losing dexterity
  • Neptune™coating on the glove fibres repel liquids, avoiding damage to the Kevlar™ Aramid fibers strands and extending glove life
  • Reinforced thumb welt increases abrasion resistance also adding to glove durability
  • Ambidextrous gloves are fully washable and can be re-used many times

Applications:

  • Chassis assembly, body trimming
  • Inspection (picking, checking and marking)
  • Metalworking and assembly operations
  • Seizing structural and body parts
  • Touching up
TRUSTED_SUSTAINABILITY

Specifications

Back Material Neptune™
Certifications/Compliance Standards CATEGORY III EN 388 : 144X, EN 407, ANSI
Cuff Material Neptune™
Coating Material Uncoated
Color Yellow
Cut Resistance Rating ANSI A2
Hand Specific
Latex Free Yes
Material Aramid Fiber
Product Type Heavyweight Gloves
Size 8
Glove Cuff Style Knit
Liner Material Dupont™ Kevlar™ aramid fibers
Lining Lined
Quantity 144 Pairs
Resistant To Very high cut resistance
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.