Learn More
Hamilton™ 700 Microliter Syringes: N Termination
The 700 series syringe is the original hand-fitted Hamilton syringe. This series of syringes is ideal for dispensing volumes from 0.5 μL up to 500 μL. A variety of different terminations and needle options are available in this series.
Brand:  Hamilton™ 87919
 View more versions of this product
Product Code. 10020332
  • £137.00 / Each
Explore more special offers
Description
Description
  • Original hand-fitted Hamilton syringe
  • Ideal for dispensing volumes from 0.5 µL up to 500 µL
  • A variety of different terminations and needle options are available in this series
Specifications
Specifications
Not Autoclavable
0.014 in.
0.13 mm
0.260 in.
6.60 mm
For dispensing low volumes to achieve the highest level of accuracy and precision possible
2 in.
Microliter Syringe
75
5 μL
26s G
0.005 in.
0.260 in.
0.343 mm
0.483 mm
2 in.
51 mm
2000 psi
3
3