Learn More
Hamilton™ 7000 Series Knurled Hub Syringes
Modified Microliter syringes with a plunger-in-needle design.
£167.00 - £1720.00
Products 4
Product Code Brand Volume (Metric) Needle Gauge Price Quantity & Availability  
Product Code Brand Volume (Metric) Needle Gauge Price Quantity & Availability  
15153038
View Documents
Hamilton™
80108
1 μL 25 G
£219.00
Each
10231432
View Documents
Hamilton™
80108
1 μL 25 G This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11518044
View Documents
Hamilton™
DS88400
£1720.00
Each
10005590
View Documents
Hamilton™
DS88500
23 G
£1720.00
Each
Description
Description
  • The only zero dead volume syringe available
  • Fine gauge tungsten plunger wire extends to the tip of the needle
  • Ensures that all solvent is expelled