missing translation for 'onlineSavingsMsg'
Learn More

Ratiolab™ 72 L BIOHAZARD Autoclavable Bags

For the sterilization and disposal of hazardous and contaminated labware.

Brand:  Ratiolab™ 7002305

Product Code. 15983730

  • £68.90 / Pack of 75

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Manufactured from very strong polypropylene film
  • With a broad seam at the bottom
  • Autoclavable at 134°C, for 20 minutes (bag must not be completely closed)
  • Rectangular white patch field turns into dark when autoclaved
  • With BIOHAZARD sticker and safety imprint in English, German, French and Spanish language
  • Provided 75 to a pack
Specifications

Specifications

Autoclavable
Single-use
Indicator patch
75/Pk.
72 L
Hazardous waste disposal
Polypropylene
700 x 1100 x 0.050 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers