Learn More
Applied Biosystems™ 7900HT TaqMan™ Low Density Array Instrument Verification RNase P Kit
Used to validate the performance of the Applied Biosystems™ 7900HT Fast Real-Time PCR Systems with the TaqMan™ Low Density Array Block Upgrade.
Brand:  Applied Biosystems™ 4351468
1019.25 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Product Code. 10005414
  • £2265.00 / Each
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
The 7900HT TaqMan™ Low Density Array Instrument Verification RNase P kit contains an empty TaqMan™ Low Density Array and 8 vials of RNase P detection reagents to detect and quantitate genomic copies of the human RNase P gene, a single-copy gene encoding the RNA moiety of the RNase P enzyme. Each vial contains premixed reagents (1X TaqMan™ Universal PCR Master Mix, RNase P primers, and FAM™ dye–labeled probe) and template. The 7900HT TaqMan™ Low Density Array Instrument Verification RNase P kit must demonstrate the ability to distinguish between 5,000 and 10,000 genomic equivalents with a 99.7% confidence level for a subsequent sample run in a single well. To ensure accurate results, Applied Biosystems™ recommends that the performance of your instrument be verified every six months.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Contains an empty TaqMan™ Low Density Array and 8 vials of RNase P detection reagents.
Spectral Calibration Kit
Fluorescent Detection
Instrument Verification Plate, RNase P Calibration
7900HT System
Real time PCR (qPCR)
1 Kit