missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ 7X Cleaning Solution

Clean test tubes, petri-dishes, flasks, cylinders, burettes, pipettes, slide cover glasses, hypodermic syringes and all other types of laboratory glassware and surgical instruments. 7X Cleaning Solution is ideal for use whenever a high degree of cleanliness is required.

Brand:  MP Biomedicals™ 0976670A4


Product Code. 15729180

  • £202.00 / Pack of 12

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Case of 12
This item is not returnable. View return policy

Description

Description

Applications: 7X cleaning solution was developed for use whenever a high degree of cleanliness is required. Test tubes, petri-dishes, flasks, cylinders, burettes, pipettes, slide cover glasses, hypodermic syringes and all other types of laboratory glassware and surgical instruments can be cleaned with 7X solution. Whenever critical cleaning procedures are involved, the use of 7X solution is indicated. 7X detergents show a broad range of application other than glassware, and have been used to clean re-usable parts of the endoscopic instruments, needles, laboratory surfaces, and stainless stain instruments and titanium components without damaging the support material. Moreover, optical components of instruments are efficiently cleaned because no fluorescent residues are left after rinsing.
Product Overview: 7X solutions are immediately and completely soluble in water at any concentration, without agitation. With normal detergents the precipitation of solids on cleaned surfaces is a major problem. The powerful dispersing agents in the 7X solutions work to reduce agglomerates to individual particles and hold them in suspension so that they are not redeposited on the surfaces being cleaned. 7X solutions contain a powerful wetting agent which will penetrate and clean minute crevices not accessible to ordinary cleaners. Laboratory equipment and glassware washed in any of the 7X solutions drain completely in a few seconds. Solutions which do not drain quickly can allow molecules of solute to be deposited on the washed surface to the extent that a visible film is present. Such deposits can affect critical experimental work. 7X cleaning solutions are compatible with alkalis and acids. The pH is near normal and varies only minimally with the concentration. The neutral pH levels will not induce etching of delicate glassware, thereby reducing weight loss and premature breakage. 7X cleaning solutions are of particular value in cell culture laboratories where any trace of chemical contamination can interfere with the growth of cell cultures in tubes or flasks.
Soluble in water at ANY concentration
Safe for glass or plastic labware
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cloudy Liquid
Cloudy Blue
Store at Room Temperature (15°C to 30°C), All 7X Cleaning Solutions have an indefinite shelf life in their original containers, at room temperature.
Case of 12
6.0 to 7.5
7X Cleaning Solutions are a concentrated liquid, made up of anionic surface active agent and special
Lab Essentials, 7X is completely soluble in water at any concentration.
16 oz.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 7X Cleaning solution

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : 2-(2-butoxyethoxy)ethanol, Dioctyl Sulfosuccinate Sodium Salt

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.