missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ 7X™ Laboratory Detergents

Safe, effective cleaning solutions for glass and plastic labware

Brand:  MP Biomedicals™ 097667098


 View more versions of this product

Product Code. 17194175

  • £1540.00 / 208.19 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

7X cleaning solution was developed for safe and effective cleaning of laboratory instruments and glassware including test tubes, petri-dishes, flasks, microscopy slides, pathology equipment and surgical instruments

  • Effective and safe
  • Completely drains from glassware in seconds
  • Eliminates residue leaving no interfering fluorescence
  • pH neutral, and no need for pH adjustment at any concentration
  • No etch to glass or plastic labware at any concentration
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Liquid
7X
Use on Glass, Plastic, benchtop equipment and tubing
6 to 7.5
15°C to 30°C
55 gal.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 7X Cleaning Solution

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : 2-(2-butoxyethoxy)ethanol, Dioctyl Sulfosuccinate Sodium Salt

SDS
Documents

Documents

Certificates
Special Offers

Special Offers