missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ 7X™-PF-O-Matic Detergent

7X™-PF-O-Matic Detergent is a Phosphate Free detergent, specifically developed for safe and effective cleaning of laboratory instruments and glassware.

Brand:  MP Biomedicals™ 097667593


 View more versions of this product

Product Code. 11479585

  • £129.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Environmentally safe and phosphate free
  • Developed for safe and effective cleaning of laboratory instruments and glassware including test tubes, petri-dishes, flasks, microscopy slides, pathology equipment and surgical instruments
  • Powerful wetting and penetrating agent, designed to clean under pressure, heat and agitation
  • Behaves and remains mostly suds-free when agitated
  • Several convenient package sizes are available
  • Stable for long-term storage
  • Completely drains from glassware in seconds
  • Eliminates residue leaving no interfering fluorescence
  • pH neutral, and no need for pH adjustment at any concentration
  • No etch to glass or plastic labware at any concentration
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Liquid
Dark Yellow
15°C to 30°C
1 gal.
7 to 8
7X
Use on Glass, Plastic, automatic washers
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • ES-7X-O-MATIC

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : sodium xylenesulphonate, polyproproxy propanol, ethylene glycol, 2,4,7,9-tetramethyldec-5-yne-4,7-di

SDS
Documents

Documents

Certificates
Special Offers

Special Offers