missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ 7X™-PF-O-Matic Detergent
7X™-PF-O-Matic Detergent is a Phosphate Free detergent, specifically developed for safe and effective cleaning of laboratory instruments and glassware.
Brand: MP Biomedicals™ 097667595
284.00 GBP valid until 2024-12-20
Use promo code "21644" to get your promotional price.
This item is not returnable.
View return policy
Description
- Environmentally safe and phosphate free
- Developed for safe and effective cleaning of laboratory instruments and glassware including test tubes, petri-dishes, flasks, microscopy slides, pathology equipment and surgical instruments
- Powerful wetting and penetrating agent, designed to clean under pressure, heat and agitation
- Behaves and remains mostly suds-free when agitated
- Several convenient package sizes are available
- Stable for long-term storage
- Completely drains from glassware in seconds
- Eliminates residue leaving no interfering fluorescence
- pH neutral, and no need for pH adjustment at any concentration
- No etch to glass or plastic labware at any concentration
Specifications
Liquid | |
Dark Yellow | |
15°C to 30°C | |
5 gal. |
7 to 8 | |
7X | |
Use on Glass, Plastic, automatic washers |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- ES-7X-O-MATIC
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H373-May cause damage to organs.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : OCTYLPHENOXYPOLYETHOXY - POLYPROPROXY PROPANOL, sodium xylenesulphonate, ethelene glycol