missing translation for 'onlineSavingsMsg'
Learn More

8-Aminonaphthalene-2-sulfonic acid, 95%, Thermo Scientific Chemicals

Product Code. 15416829
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-28-8
Molecular Formula C10H9NO3S
Molecular Weight (g/mol) 223.246
MDL Number MFCD00004032
InChI Key QEZZCWMQXHXAFG-UHFFFAOYSA-N
Synonym 8-amino-2-naphthalenesulfonic acid, 1,7-cleve's acid, 1-naphthylamine-7-sulfonic acid, 2-naphthalenesulfonic acid, 8-amino, cleve's theta-acid, 1-amino-7-sulfonaphthalene, 1-aminonaphthalene-7-sulfonic acid, 8-naphthylamine-2-sulfonic acid, 1-amino-7-naphthalenesulfonic acid, 1-amino-naphthalene-7-sulphonic acid
PubChem CID 8386
IUPAC Name 8-aminonaphthalene-2-sulfonic acid
SMILES C1=CC2=C(C=C(C=C2)S(=O)(=O)O)C(=C1)N

Specifications

Melting Point >300°C
Color Gray to Yellow
Quantity 25 g
UN Number UN2585
Beilstein 2115629
Merck Index 142,351
Formula Weight 223.25
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 8-Aminonaphthalene-2-sulfonic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'