missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Bromo-2'-deoxyguanosine, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
Description
8-Bromo-2'-deoxyguanosine is useful as the starting structure for 2'-deoxyguanosine nucleotides, which are modified in position 8.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
8-Bromo-2′-deoxyguanosine is useful as the starting structure for 2′-deoxyguanosine nucleotides, which are modified in position 8.
Solubility
Soluble in dimethyl sulfoxide, warm water and dimethyl formam
8-Bromo-2′-deoxyguanosine is useful as the starting structure for 2′-deoxyguanosine nucleotides, which are modified in position 8.
Solubility
Soluble in dimethyl sulfoxide, warm water and dimethyl formam
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 13389-03-2 |
Melting Point | >250°C (decomposition) |
Color | White |
Molecular Formula | C10H12BrN5O4 |
MDL Number | MFCD01630971 |
Quantity | 1 g |
Synonym | 8-bromo-2'-deoxyguanosine, 8-bromo-2'-dg, 2-amino-8-bromo-9-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-1h-purin-6 9h-one, 8-br-2'-dg, guanosine, 8-bromo-2'-deoxy, 8-bromodeoxyguanosine, epitope id:151524, 8-bromo-2?-deoxyguanosine, guanosine,8-bromo-2'-deoxy, 8-bromo-2-deoxyguanosine |
Solubility Information | Soluble in dimethyl sulfoxide,warm water and dimethyl formamide. |
InChI Key | MKDXZFVCXWXGBQ-QXZWWJJYNA-N |
SMILES | NC1=NC2=C(N=C(Br)N2[C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N1 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'