missing translation for 'onlineSavingsMsg'
Learn More

8-Bromo-6-chloroimidazo[1,2-a]pyridine, 95%, Thermo Scientific Chemicals

Product Code. 15407467
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

8-Bromo-6-chloroimidazo[1,2-a]pyridine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Bromo-6-chloroimidazo[1,2-a]pyridine is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 957187-27-8
Molecular Formula C7H4BrClN2
Molecular Weight (g/mol) 231.48
MDL Number MFCD12828062
InChI Key LMPPBTNPACXNDI-UHFFFAOYSA-N
Synonym 8-bromo-6-chloroimidazo 1,2-a pyridine, 8-bromo-6-chloro-imidazo 1,2-a pyridine, 6-chloro-8-bromoimidazo 1,2-a pyridine
PubChem CID 26370038
IUPAC Name 8-bromo-6-chloroimidazo[1,2-a]pyridine
SMILES ClC1=CN2C=CN=C2C(Br)=C1

Specifications

Melting Point 125°C to 128°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 231.48
Percent Purity 95%
Chemical Name or Material 8-Bromo-6-chloroimidazo[1,2-a]pyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.