missing translation for 'onlineSavingsMsg'
Learn More

8-Bromoadenosine-3',5'-cyclic monophosphate, Thermo Scientific Chemicals

Product Code. 15426409
50 mg
Click to view available options
Quantity:
50 mg
100 mg
250 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23583-48-4
Molecular Formula C10H11BrN5O6P
Molecular Weight (g/mol) 408.11
MDL Number MFCD00075580
InChI Key DVKQVRZMKBDMDH-UUOKFMHZSA-N
Synonym bcamp, 8-bromo-cyclic amp, 8-br-camp, 8-bromoadenosine 3',5'-cyclic monophosphate, 8-bromo-cyclic 3',5'-amp, 8-bromo-camp, 8-bromo cyclic adenosine monophosphate, 8-bromoadenosine 3',5'-monophosphate, 8-bromoadenosine cyclic 3',5'-phosphate, cyclic 8-bromoadenosine 3',5'-monophosphateShow MoreShow Less
PubChem CID 32014
ChEBI CHEBI:64211
IUPAC Name (4aR,6R,7R,7aS)-6-(6-amino-8-bromopurin-9-yl)-2-hydroxy-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-7-ol
SMILES NC1=C2N=C(Br)N([C@@H]3O[C@@H]4COP(O)(=O)O[C@H]4[C@H]3O)C2=NC=N1

Specifications

Melting Point 254°C
Color Brown
Quantity 50 mg
Sensitivity Hygroscopic
Formula Weight 408.11
Physical Form Powder
Chemical Name or Material 8-Bromoadenosine-3',5'-cyclic monophosphate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU27K044 Certificate TypeCertificate of Analysis Date05/10/2024 Catalog NumberJ64469.MF,J64469.MD,J64469.MC
Lot NumberS19K014 Certificate TypeCertificate of Analysis Date01/07/2024 Catalog NumberJ64469.MF,J64469.MD,J64469.MC
Lot NumberQ23K027 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ64469.MF,J64469.MD,J64469.MC
Lot NumberN05K027 Certificate TypeCertificate of Analysis Date16/02/2024 Catalog NumberJ64469.MF,J64469.MD,J64469.MC
Lot NumberM22K012 Certificate TypeCertificate of Analysis Date26/01/2024 Catalog NumberJ64469.MF,J64469.MD,J64469.MC

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
8-Bromoadenosine-3',5'-cyclic monophosphate, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.