missing translation for 'onlineSavingsMsg'
Learn More

8-Bromoimidazo[1,2-a]pyridine, 95%, Thermo Scientific Chemicals

Product Code. 12307183
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850349-02-9
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.03
InChI Key GCCVNNZARPFXES-UHFFFAOYSA-N
PubChem CID 17749933
IUPAC Name 8-bromoimidazo[1,2-a]pyridine
SMILES C1=CN2C=CN=C2C(=C1)Br

Specifications

Melting Point 72°C to 75°C
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Synonym 8-bromoimidazo 1,2-a pyridine, 8-bromo-imidazo 1,2-a pyridine, 8-bromoh-imidazo 1,2-a pyridine, imidazo 1,2-a pyridine,8-bromo, imidazo 1,2-a pyridine, 8-bromo
Formula Weight 197.03
Percent Purity 95%
Chemical Name or Material 8-Bromoimidazo[1, 2-a]pyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'