Learn More
8-Bromoimidazo[1,2-a]pyridine, 95%, Thermo Scientific Chemicals
£90.60 - £363.97
Chemical Identifiers
CAS | 850349-02-9 |
---|---|
Molecular Formula | C7H5BrN2 |
Molecular Weight (g/mol) | 197.03 |
InChI Key | GCCVNNZARPFXES-UHFFFAOYSA-N |
PubChem CID | 17749933 |
IUPAC Name | 8-bromoimidazo[1,2-a]pyridine |
SMILES | C1=CN2C=CN=C2C(=C1)Br |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
850349-02-9 | |
197.03 | |
17749933 | |
C1=CN2C=CN=C2C(=C1)Br |
C7H5BrN2 | |
GCCVNNZARPFXES-UHFFFAOYSA-N | |
8-bromoimidazo[1,2-a]pyridine |
Specifications
850349-02-9 | |
94% min. (GC) | |
C7H5BrN2 | |
GCCVNNZARPFXES-UHFFFAOYSA-N | |
8-bromoimidazo[1,2-a]pyridine | |
17749933 | |
95% |
72°C to 75°C | |
Glass Bottle | |
8-bromoimidazo 1,2-a pyridine, 8-bromo-imidazo 1,2-a pyridine, 8-bromoh-imidazo 1,2-a pyridine, imidazo 1,2-a pyridine,8-bromo, imidazo 1,2-a pyridine, 8-bromo | |
C1=CN2C=CN=C2C(=C1)Br | |
197.03 | |
197.03 | |
8-Bromoimidazo[1, 2-a]pyridine |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and ke
GHS Signal Word: Warning
RUO – Research Use Only