missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ 3500 Genetic Analyzer 8-Capillary Array, 36 cm
A radio-frequency identification tag is built into the frame for tracking critical run information, which is then reported on the software dashboard.
Brand: Applied Biosystems™ 4404683
This item is not returnable.
View return policy
Description
The internally uncoated capillaries are supplied in preassembled sets of 8 capillaries per array, with a built-in frame for easy installation.Order Info
Each
Specifications
36 cm | |
3500 Genetic Analyzer 8-Capillary Array, 36cm Capillary, Internally Uncoated Capillaries, For Use With 3500 Genetic Analyzer, 15 to 30°C Storage Temperature | |
3500 Genetic Analyzer | |
Genetic Analyzer 8-Capillary Array | |
Room Temperature |
Store this array at ambient temperature. For details about storing the capillary array on and off the instrument, please refer to the Applied Biosystems 3500/3500xL Genetic Analyzer User Guide. | |
Human Identification, Sequencing, Fragment Analysis | |
36 cm (Capillary) | |
1 array |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction