Learn More
Integra Biosciences™ 8-channel Adapter
Use these adapters with VACUBOY aspiration tools. Integra Biosciences™ 8-Channel Adapters used with VACUBOY aspiration tools convert any vacuum source into a user-friendly aspiration system.
£81.89 - £114.00
Specifications
For Use With (Equipment) VACUBOY Aspiration Tools
View More Specs
Products 2
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
10023451
View Documents
Integra Biosciences™
155503
8-Channel stainless steel adapter, 40mm
£114.00
Each
In Stock
to see stock.
Edge
Add to basket
12603275
View Documents
Integra Biosciences™
159025
8-Channel adapter with ejector for grip tips
£81.89
Each
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
Description
Description
  • 8-Channel adapter
  • Flame sterilizable for maximal safety
  • Fit a variety of tubes, flasks, bottles etc.
  • Accommodate 12- to 96-well plates
Can be used with VACUSAFE comfort vacuum system. Enables liquid residues to be disposed of reliably, precisely and conveniently:
  • With a precisely dosed volume
  • Without loss of drops
  • Without risk of contamination
  • Without causing tiredness
  • All parts in contact with fluids are autoclavable
Specifications
Specifications
VACUBOY Aspiration Tools
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Integra Biosciences™ 8-channel Adapter