Learn More
Cytiva Whatman™ 8-Channel Filter Plates
For use with Agilent 850-DS dissolution sampling station
Brand:  Cytiva 7707-3500
Code : NEW
Additional Details : Weight : 1.71000kg
 View more versions of this product
Product Code. 15528753
  • £545.00 / Pack of 50
Estimated Shipment: 28-10-2024
to see stock.
Add to basket
Description
Description
  • Available in PTFE, Nylon, PES, and GMF filter media
  • Disposable
  • Automated dissolution testing
Specifications
Specifications
Agilent 850-DS Dissolution Sampling Station
Nylon
0.2 μm
Automated dissolution testing