missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 8-Hydroxy-1,3,6-pyrenetrisulfonic acid trisodium salt, 98%

Product Code. 10445005
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6358-69-6
Molecular Formula C16H7Na3O10S3
Molecular Weight (g/mol) 524.37
MDL Number MFCD00037575
InChI Key KXXXUIKPSVVSAW-UHFFFAOYSA-K
PubChem CID 61388
ChEBI CHEBI:52083
IUPAC Name trisodium;8-hydroxypyrene-1,3,6-trisulfonate
SMILES [Na+].[Na+].[Na+].OC1=CC(=C2C=CC3=C(C=C(C4=CC=C1C2=C34)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Green-Yellow to Yellow
Density 2.1500g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 8065
Specific Gravity 2.15
Synonym pyranine, solvent green 7, c.i. solvent green 7, japan green 204, hpts, FD&C green no. 8, japan green no. 204, pyrene 1, pyranine 120, FD&C green 8
Solubility Information Solubility in water: 300g/L (25°C).
Formula Weight 524.39
Percent Purity 98%
Grade Extra Pure
Physical Form Crystals or Powder
Chemical Name or Material 8-Hydroxy-1, 3, 6-pyrenetrisulfonic acid trisodium salt
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.