missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Hydroxyquinoline-5-sulfonic acid monohydrate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | >300.0°C |
Color | Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min |
Packaging | Plastic bottle |
Molecular Formula | C9H7NO4S·H2O |
Quantity | 2.5 kg |
Beilstein | 22,407 |
Merck Index | 14,4844 |
Solubility Information | Solubility in water: freely soluble. Other solubilities: slightly soluble in most common organic solvents |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction