Learn More
8-Hydroxyquinoline sulfate monohydrate, 98%, Thermo Scientific Chemicals
£29.60 - £650.00
Chemical Identifiers
CAS | 207386-91-2 |
---|---|
Molecular Formula | 0·5 H2O |
Molecular Weight (g/mol) | 203.2 |
MDL Number | MFCD00149610 |
InChI Key | BNCXJZDIJIVJJO-UHFFFAOYSA-N |
Synonym | 8-quinolinol hemisulfate, 8-quinolinol hemisulfate hemihydrate, 8-hydroxyquinoline hemisulfate salt hemihydrate, quinolin-8-ol; sulfuric acid; hydrate, quinolin-8-ol sulfate hydrate 2:1:1, 8-hydroxyquinoline sulfate, 8-quinolinol sulfate 2:1 monohydrate, 8-quinolinol, sulfate, hydrate 2:1:1, 8-quinolinol hemisulfate salt hemihydrate, pestanal r , analytical standard |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10665002
|
Thermo Scientific Acros
148451000 |
100 g | Glass bottle |
£29.60
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
10533121
|
Thermo Scientific Acros
148455000 |
500 g | Glass bottle |
£117.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10237113
|
Thermo Scientific Acros
148450025 |
2.5 kg | Plastic Bottle |
£650.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
207386-91-2 | |
203.2 | |
BNCXJZDIJIVJJO-UHFFFAOYSA-N |
0·5 H2O | |
MFCD00149610 | |
8-quinolinol hemisulfate, 8-quinolinol hemisulfate hemihydrate, 8-hydroxyquinoline hemisulfate salt hemihydrate, quinolin-8-ol; sulfuric acid; hydrate, quinolin-8-ol sulfate hydrate 2:1:1, 8-hydroxyquinoline sulfate, 8-quinolinol sulfate 2:1 monohydrate, 8-quinolinol, sulfate, hydrate 2:1:1, 8-quinolinol hemisulfate salt hemihydrate, pestanal r , analytical standard |
Specifications
207386-91-2 | |
100.0 | |
Yellow | |
98% | |
MFCD00149610 | |
8-quinolinol hemisulfate, 8-quinolinol hemisulfate hemihydrate, 8-hydroxyquinoline hemisulfate salt hemihydrate, quinolin-8-ol; sulfuric acid; hydrate, quinolin-8-ol sulfate hydrate 2:1:1, 8-hydroxyquinoline sulfate, 8-quinolinol sulfate 2:1 monohydrate, 8-quinolinol, sulfate, hydrate 2:1:1, 8-quinolinol hemisulfate salt hemihydrate, pestanal r , analytical standard | |
BNCXJZDIJIVJJO-UHFFFAOYSA-N | |
203.2 | |
Crystalline Powder or Lumps |
97.5 | |
182.0°C | |
Authentic | |
0·5 H2O | |
14, 4843 | |
Solubility in water: soluble in water. Other solubilities: slightly soluble in alcohol, insoluble in ether | |
203.2 | |
98% | |
8-Hydroxyquinoline sulfate monohydrate, 98% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
- 8-Hydroxyquinoline sulfate monohydrate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only