missing translation for 'onlineSavingsMsg'
Learn More

8-Methyl-8-azabicyclo[3.2.1]octan-3-amine dihydrochloride, 90%, Thermo Scientific™

£31.10 - £225.00

Chemical Identifiers

CAS 646477-45-4
Molecular Formula C8H18Cl2N2
Molecular Weight (g/mol) 213.146
MDL Number MFCD07366481
InChI Key KVYQKWPZQKGICY-UHFFFAOYSA-N
Synonym 8-methyl-8-azabicyclo 3.2.1 octan-3-amine dihydrochloride, 8-methyl-8-aza-bicyclo 3.2.1 octan-3-amine dihydrochloride, 8-azabicyclo 3.2.1 octan-3-amine, 8-methyl-, dihydrochloride, 3-alpha-aminotropane dihydrochloride, 8-methyl-8-azabicyclo 3.2.1 oct-3-ylamine dihydrochloride, 3-amino-8-methyl-8-azabicyclo 3.2.1 octane dihydrochloride, 8-methyl-8-aza-bicyclo 3.2.1 octan-3-aminedihydrochloride
PubChem CID 2775928
IUPAC Name 8-methyl-8-azabicyclo[3.2.1]octan-3-amine;dihydrochloride
SMILES CN1C2CCC1CC(C2)N.Cl.Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10314272
View Documents
Thermo Scientific Maybridge
BTB14320DA
1 g Amber Glass Bottle
£31.10
1g
Please to purchase this item. Need a web account? Register with us today!
10374362
View Documents
Thermo Scientific Maybridge
BTB14320EA
10 g Amber Glass Bottle
£225.00
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

646477-45-4
213.146
KVYQKWPZQKGICY-UHFFFAOYSA-N
2775928
CN1C2CCC1CC(C2)N.Cl.Cl
C8H18Cl2N2
MFCD07366481
8-methyl-8-azabicyclo 3.2.1 octan-3-amine dihydrochloride, 8-methyl-8-aza-bicyclo 3.2.1 octan-3-amine dihydrochloride, 8-azabicyclo 3.2.1 octan-3-amine, 8-methyl-, dihydrochloride, 3-alpha-aminotropane dihydrochloride, 8-methyl-8-azabicyclo 3.2.1 oct-3-ylamine dihydrochloride, 3-amino-8-methyl-8-azabicyclo 3.2.1 octane dihydrochloride, 8-methyl-8-aza-bicyclo 3.2.1 octan-3-aminedihydrochloride
8-methyl-8-azabicyclo[3.2.1]octan-3-amine;dihydrochloride

Specifications

646477-45-4
100.0
C8H18Cl2N2
8-methyl-8-azabicyclo 3.2.1 octan-3-amine dihydrochloride, 8-methyl-8-aza-bicyclo 3.2.1 octan-3-amine dihydrochloride, 8-azabicyclo 3.2.1 octan-3-amine, 8-methyl-, dihydrochloride, 3-alpha-aminotropane dihydrochloride, 8-methyl-8-azabicyclo 3.2.1 oct-3-ylamine dihydrochloride, 3-amino-8-methyl-8-azabicyclo 3.2.1 octane dihydrochloride, 8-methyl-8-aza-bicyclo 3.2.1 octan-3-aminedihydrochloride
CN1C2CCC1CC(C2)N.Cl.Cl
213.146
213.15
8-Methyl-8-azabicyclo[3.2.1]octan-3-amine dihydrochloride
89.5
90%
MFCD07366481
KVYQKWPZQKGICY-UHFFFAOYSA-N
8-methyl-8-azabicyclo[3.2.1]octan-3-amine;dihydrochloride
2775928
90%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
Documents

Documents

Product Certifications