missing translation for 'onlineSavingsMsg'
Learn More

8-Octanolactam, 98%, Thermo Scientific Chemicals

Product Code. 11438557
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Azacyclononanone has been used as an internal standard liquid chromatography-ultraviolet detection method for the determination of caprolactam in the aqueous food stimulant 3% acetic acid (w/v).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Azacyclononanone has been used as an internal standard liquid chromatography-ultraviolet detection method for the determination of caprolactam in the aqueous food stimulant 3% acetic acid (w/v).

Solubility
Soluble in methanol (almost transparency).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 935-30-8
Molecular Formula C8H15NO
Molecular Weight (g/mol) 141.214
MDL Number MFCD00003273
InChI Key YDLSUFFXJYEVHW-UHFFFAOYSA-N
Synonym 2-azacyclononanone, caprylolactam, capryllactam, azacyclononan-2-one, 8-octanelactam, 8-aminooctanoic acid lactam, 2-perhydroazoninone, 2h-azonin-2-one, octahydro, cyclooctanone lactam, octamethylenimine, 2-oxo
PubChem CID 13632
IUPAC Name azonan-2-one
SMILES C1CCCC(=O)NCCC1

Specifications

Melting Point 72°C to 75°C
Quantity 25 g
Beilstein 109775
Solubility Information Soluble in methanol (almost transparency).
Formula Weight 141.21
Percent Purity 98%
Chemical Name or Material 8-Octanolactam

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'