missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 8 x 50mL Fixed Angle Rotor

Carry out high-risk applications with the Thermo Scientific™ 8 x 50mL Fixed-Angle Rotor.

Brand:  Thermo Scientific™ 75003335

Product Code. 10543812

  • £2551.25 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Carry out high-risk applications with the Thermo Scientific™ 8 x 50mL Fixed-Angle Rotor.

Ordering Information:

Order rotor body (75-003-335) and components to complete assembly. Order 75-003-404 for O-ring/gasket replacement. Tube adapters are also available separately. Contact your sales representative for information.

Compatible with:

Thermo Scientific™ Stratos Biofuge, Sorvall™ Stratos centrifuge series

TRUSTED_SUSTAINABILITY
Specifications

Specifications

8 x 50mL Fixed Angle Rotor
8 x 50 mL
13,000 rpm (15,000 rpm 208V)
5.6 cm
Thermo Scientific Sorvall Stratos and Stratos Biofuge Centrifuges
Biocontainment Certified by CAMR™, HPA, Porton Down, U.K.
Fixed Angle Rotor
20,220 x g (26,920 x g with 208V )
10.7 cm
971 (For 120V unit)
729 (For 208V unit)
34°
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers