missing translation for 'onlineSavingsMsg'
Learn More
Learn More
80% Ethanol, Molecular Biology Grade, Thermo Scientific™
Ready made ethanol/water blend prequalified for molecular biology research applications.
£149.00 - £188.00
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
16649582
|
Thermo Scientific
T08204DF2500 |
2.5 L | Glass Bottle |
£149.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16622284
|
Fisher Chemical
T08204K7 |
4 L | Poly Bottle |
£188.00
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- 80% ethanol and water blend
- DNase-, RNase-, and protease-free
- Ready made ethanol/water blend
- Pre-qualified for molecular biology research applications
- Suitable for the purification and precipitation of nucleic acids (DNA and RNA) and proteins
- Tested for use with Applied Biosystems™ MagMAX™ kits
Chemical Identifiers
64-17-5, 7732-18-5 |
Specifications
64-17-5, 7732-18-5 | |
78% to 82% | |
80% | |
Ethyl Alcohol |
Clear, Colorless Liquid | |
0.001% Maximum | |
Molecular Biology |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 80% Ethanol, for Molecular Biology, Duty Free
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P370+P378-In case of fire: Use to extinguish.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
For Manufacturing and Laboratory Use. Not for beverage use.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
80% Ethanol, Molecular Biology Grade, Thermo Scientific™