missing translation for 'onlineSavingsMsg'
Learn More
Learn More
80% Ethanol, Molecular Biology Grade, Thermo Scientific™
Click to view available options
2.5 L; Glass Bottle
Description
- 80% ethanol and water blend
- DNase-, RNase-, and protease-free
- Ready made ethanol/water blend
- Pre-qualified for molecular biology research applications
- Suitable for the purification and precipitation of nucleic acids (DNA and RNA) and proteins
- Tested for use with Applied Biosystems™ MagMAX™ kits
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 64-17-5, 7732-18-5 |
Color | Clear, Colorless Liquid |
Assay Percent Range | 78% to 82% |
Molecular Formula | C2H6O |
InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
SMILES | CCO |
IUPAC Name | ethanol |
Molecular Weight (g/mol) | 46.07 |
Percent Purity | 80% |
Grade | Molecular Biology |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'